pH combination electrodes, IDS®
Toimittaja: SI Analytics
285100080 285100120 285101080
SCOI285100080EA 733 EUR
SCOI285100080 661-0244 SCOI285101080
pH combination electrodes, IDS®
Elektrodit ja niiden oheistarvikkeet pH-elektrodit
When digital IDS sensors are connected to the measuring device, they automatically identify themselves with their serial numbers and type designation and transmit their calibration data to the device.
  • Calibration data is stored in the sensor head
  • Measuring signals are transmitted without interference
  • Perfect galvanized separation
  • Resistant against environmental influences
  • Allows prognostic maintenance of the sensors by the intelligent sensor evaluation QSC
  • Effortless allocation and documentation of the sensor to electronically captured and saved