Etikkahappo 75% ≥75% for biopharmaceutical production

Toimittaja: VWR Chemicals
Vaara

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

6519.2500EA 80.1 EUR
6519.2500 6519.9025
Etikkahappo 75% ≥75% for biopharmaceutical production
Etikkahappo
Kaava: H₃CCOOH
MW: 60.05 g/mol
Kiehumispiste: 118
Sulamispiste: 17
Leimahduspiste: 38.5
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00036152
CAS-numero: 64-19-7
EINECS: 200-580-7
UN: 2790
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR