Anti-SELL Rat Monoclonal Antibody (PerCP-Cyanine5.5) [clone: MEL-14]
Toimittaja: Tonbo Biosciences
65-0621-U025 65-0621-U100
65-0621-U025EA 104 EUR
65-0621-U025 65-0621-U100
Anti-SELL Rat Monoclonal Antibody (PerCP-Cyanine5.5) [clone: MEL-14]
Vasta-aineet
The MEL-14 antibody is specific for mouse CD62L, also known as L-Selectin, a cell adhesion molecule which facilitates lymphocyte "rolling" on activated vascular endothelium and homing to high endothelial venules (HEV) as immune cells transmigrate from blood into peripheral tissues. L-Selectin is a member of a family of Selectin molecules which act together with the integrin family of adhesion molecules to mediate leukocyte-endothelial interactions. L-Selectin is characteristically expressed by neutrophils, and is also found on B cells, monocytes, granulocytes, and at varying levels on naive,More Product Information