Anti-CD8A Mouse Monoclonal Antibody (Peridinin Chlorophyll/Cy5.5®) [clone: RPA-T8]
Toimittaja: Tonbo Biosciences
65-0088-T025 65-0088-T100
65-0088-T025EA 90.6 EUR
65-0088-T025 65-0088-T100
Anti-CD8A Mouse Monoclonal Antibody (Peridinin Chlorophyll/Cy5.5®) [clone: RPA-T8]
Vasta-aineet
The RPA-T8 antibody is specific for the 32-34 kDa alpha chain of human CD8, known as CD8a or CD8 alpha. CD8a can form a homodimer (CD8 alpha-alpha), but is more commonly expressed as a heterodimer with a second chain known as CD8b or CD8 beta. CD8 acts as a co-receptor for antigen recognition and subsequent T cell activation that is initiated upon binding of the T cell receptor (TCR) to antigen-bearing MHC Class I molecules. The cytoplasmic domains of CD8 provide binding sites for the tyrosine kinase lck, facilitating intracellular signaling events that lead to T cell activation, development,More Product Information