Fosforihappo ≥85% ≥85%, TraceSELECT™ Ultra for ultratrace analysis, Fluka™
Toimittaja: Honeywell Chemicals
|
Vaara
|
Synonyms:
Phosphoric acid
64957-1L
64957-1LEA
2810
EUR
64957-1L
Fosforihappo ≥85% ≥85%, TraceSELECT™ Ultra for ultratrace analysis, Fluka™
Fosforihappo
TraceSELECT™ Ultra and TraceSELECT
- Ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- Sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- The acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (µg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions
Kaava:
H₃PO₄ MW: 98 g/mol Kiehumispiste: 158 °C (1.013 hPa) Sulamispiste: 21 °C Tiheys: 1.71 g/cm³ (20 °C) Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD00011340 CAS-numero: 7664-38-2 EINECS: 231-633-2 UN: 1805 ADR: 8 III Merck Index: 14,07344 |
Testitulosten erittely
Assay | 85.0 - 90.0 % |
Density (D 20/4) | 1.691 - 1.706 |
Silver (Ag) | Max. 1 ppb |
Aluminium (Al) | Max. 10 ppb |
Arsenic (As) | Max. 2 ppb |
Gold (Au) | Max. 1 ppb |
Boron (B) | Max. 10 ppb |
Barium (Ba) | Max. 1 ppb |
Beryllium (Be) | Max. 5 ppb |
Bismuth (Bi) | Max. 1 ppb |
Calcium (Ca) | Max. 50 ppb |
Cadmium (Cd) | Max. 1 ppb |
Cerium (Ce) | Max. 0.5 ppb |
Cobalt (Co) | Max. 1 ppb |
Chromium (Cr) | Max. 20 ppb |
Cesium (Cs) | Max. 0.5 ppb |
Copper (Cu) | Max. 5 ppb |
Dysprosium (Dy) | Max. 0.5 ppb |
Erbium (Er) | Max. 0.5 ppb |
Europium (Eu) | Max. 0.5 ppb |
Iron (Fe) | Max. 100 ppb |
Gallium (Ga) | Max. 0.5 ppb |
Gadolinium (Gd) | Max. 1 ppb |
Germanium (Ge) | Max. 1 ppb |
Hafnium (Hf) | Max. 1 ppb |
Mercury (Hg) | Max. 0.5 ppb |
Holmium (Ho) | Max. 0.5 ppb |
Indium (In) | Max. 0.5 ppb |
Iridium (Ir) | Max. 0.5 ppb |
Potassium (K) | Max. 10 ppb |
Lanthanum (La) | Max. 0.5 ppb |
Lithium (Li) | Max. 1 ppb |
Lutetium (Lu) | Max. 0.5 ppb |
Magnesium (Mg) | Max. 5 ppb |
Manganese (Mn) | Max. 5 ppb |
Molybdenum (Mo) | Max. 5 ppb |
Sodium (Na) | Max. 50 ppb |
Niobium (nb) | Max. 5 ppb |
Neodymium (nd) | Max. 0.5 ppb |
Nickel (Ni) | Max. 20 ppb |
Lead (Pb) | Max. 1 ppb |
Palladium (Pd) | Max. 1 ppb |
Praseodymium (Pr) | Max. 0.5 ppb |
Platinum (Pt) | Max. 1 ppb |
Rubidium (Rb) | Max. 0.5 ppb |
Rhenium (Re) | Max. 0.5 ppb |
Rhodium (Rh) | Max. 0.5 ppb |
Ruthenium (Ru) | Max. 0.5 ppb |
Antimony (Sb) | Max. 500 ppb |
Selenium (Se) | Max. 5 ppb |
Samarium (Sm) | Max. 0.5 ppb |
Tin (Sn) | Max. 2 ppb |
Strontium (Sr) | Max. 1 ppb |
Tantalum (Ta) | Max. 5 ppb |
Terbium (Tb) | Max. 0.5 ppb |
Tellurium (Te) | Max. 1 ppb |
Thorium (Th) | Max. 0.5 ppb |
Titanium (Ti) | Max. 10 ppb |
Thallium (Tl) | Max. 0.5 ppb |
Thulium (Tm) | Max. 0.5 ppb |
Uran (U) | Max. 0.5 ppb |
Vanadium (V) | Max. 1 ppb |
Tungsten (W) | Max. 5 ppb |
Yttrium (Y) | Max. 0.5 ppb |
Ytterbium (Yb) | Max. 0.5 ppb |
Zinc (Zn) | Max. 10 ppb |
Zirconium (Zr) | Max. 5 ppb |
Chloride (Cl) | Max. 0.5 ppm |
Nitrate (NO₃) | Max. 0.5 ppm |
Sulfate (SO₄) | Max. 5 ppm |
Ammonium (NH₄) | Max. 1 ppm |
KMnO₄ reducing matter (as O) | Max. 1 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...