Silikageeli pore size 60 Å, 200-425 mesh particle size, Technical Grade, Supelco®

Toimittaja: Merck

645524-2.5KG 645524-5KG
645524-2.5KGEA 495 EUR
645524-2.5KG 645524-5KG
Silikageeli pore size 60 Å, 200-425 mesh particle size, Technical Grade, Supelco®
Silikageeli
Kaava: SiO₂
Kiehumispiste: 2230 °C
Sulamispiste: > 1600 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011232
CAS-numero: 63231-67-4
EINECS: 231-545-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ca78d31ff20753ad4fe555a9a4aff389df4c346&vid=f1d60a6c031c3a28813381e57c4e18551eef34ef&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd22e7ab466a8ebd53ccd6e783f7e8786f0dcc5b&vid=1b64f951cb6cf627252073c9932d4c2bc0cc6d47&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR