VWR® Culture Media, Ready To Use, In Tubes
Toimittaja: VWR Chemicals
AX011101EA 154 EUR
AX011101 AX011137 AX011120 AX011112 AX011122 AX011113 AX011124 AX011103 AX011129 AX011130 AX011139 AX011141 AX011144 AX011146 AX011155 AX011158 AX011159 710-0379 AX011176 AX011184 AX011186 351430XF 351430ZF 790360WF 361430XF 730731ZA 613270ZA 730733IA 499127OO 601320ZA 601370ZA 601410ZF 601450ZA 601580ZA 610173ZA 610753ZA 611121ZA 611314ZA 611585ZA 611830ZA 613155ZA 621051ZA 600113ZA 600343ZF 610360ZF 600834ZA 600954ZA 601083ZA 601314ZA 601320XA 640431ZA 640504ZA 690042ZA 690144ZA 690450ZA 710360ZF 721115TA 730774ZA 761320WA 731203ZA 731250ZA AX011194 AX011192 611770ZA 751121ZA 641114ZA AX011196 640971ZA 641071ZA 600304ZA 600531ZA 600592ZA 600894ZA 600944ZA 601121ZA 601770ZA 602524ZF 632100ZF 640511ZA 640884CA 651820ZA 661161XF 690440ZA 730144WA 890450WA 690674ZA 613120ZA 601121OA 690592ZA 651810ZA 601060AA 601060ZA 603552ZA 650450ZA 601030ZA 690450XA 640504XA 603572ZA 499128OO 663650YF 720582ZA 720774ZA 723650YF 640884DA 611122OA 602600OA AX011197 AX011199 AX011200 600621ZA 622370XA 730884CA 731114ZA 731183ZA 721112ZA 601850ZA 734702IA 825262ZA 825290ZA 601780ZA 745470ZF 601314ZAMP 640431ZAMP 632100ZFMP 613155ZAMP 611585ZAMP 610360ZFMP 603552ZAMP 640511ZAMP 601320XAMP 640884DAMP 601060AAMP 600954ZAMP 600834ZAMP 600621ZAMP 600531ZAMP 600343ZFMP 600304ZAMP 601370ZAMP 723650YFMP 890450WAMP 790360WFMP 761320WAMP 751121ZAMP 731250ZAMP 730884CAMP 730774ZAMP 730733IAMP 640504ZAMP 730144WAMP 721115TAMP 720582ZAMP 710360ZFMP 690674ZAMP 690450ZAMP 690450XAMP 690144ZAMP 690042ZAMP 663650YFMP 640971ZAMP 730731ZAMP 661161XFMP 651820ZAMP 733552ZA 632512ZA 825490ZA 601120AF 601314TH 651812ZA 615870ZA 610173ZAMP AX011202 AX011203 AX011204 AX011205 826030ZA 640884ZA 710884CA 616170ZA 690894ZA 705870YA 695640RA 695640XA 692260ZA 613430ZA 611310TA 640082ZA 611780ZA 616350SA 361430ZF 606460ZA 615650ZA 610343VF 690451ZA 600170ZA 612760TA 614990TA 640504AA 616620XA 601122ZA 611122ZA 725682ZA
VWR® Culture Media, Ready To Use, In Tubes
Elatusaineet mikrobiologiaan Valmisteltu viljelyaine
Erilaisiin mikrobiologian käyttötarkoituksiin.
  • Storage at room temperature for selected media
  • Compact packaging, tubes are fitted with aluminium screw caps
  • Media, batch number and expiry date printed on every single tube
Pakkaus: Supplied in boxes of 20, 50 or 120 tubes.