Lisävarusteet, Kvartsi-kontrollilevyt
Toimittaja: BELLINGHAM STANLEY
34-21 34-20 34-241 34-22 90-803
635-0023EA 1990 EUR
635-0023 635-0022 635-0666 635-0405 BELL90-803
Lisävarusteet, Kvartsi-kontrollilevyt
Standardiaineet ja vertailumateriaalit Taitekerroinstandardit
Quartz control plates for the calibration and verification of polarimeters. of optical and digital polarimeters and saccharimeters.
  • Inert quartz with relatively low temperature coefficient, easy to handle, and infinite shelf life
  • Available in three optical rotation values
  • Three decimal place uncertainty
The optional temperature saddles may used as an interface between the quartz plate and instrument temperature sensor to provide fine compensation.
Sertifikaatit: Traceable to PTB. UKAS-MRA calibration in accordance with ISO 17025:2005 between 325-880nm available.
Tilaustiedot: Sadlles can be ordered in option.
Toimitustiedot: Supplied with a certificate of verification in a plastic storage case.