Optical Elements for Special Applications
Toimittaja: HELLMA
013-000-71 013-101-71
634-9344EA 138 EUR
634-9344 634-9347
Optical Elements for Special Applications
Kyvetit Spektrofotometrin kyvetit
Hellma® precision cells are 100% manufactured in Germany and are suitable for all common spectrophotometers.
  • Holder for cuvettes with detachable windows
  • Aluminium spacer to fit cells with 1 mm optical path length into 10 mm cell holder
Highest precision: Hellma® precision cells are characterized by the highest precision in path length accuracy and parallelism. This guarantees continuously reproducible measurement results.
Chemically resistant and thermally bonded: Hellma® precision cells are thermally bonded and not glued, which gives them a very