Cole-Parmer® SP-500 Visible and UV/Visible Scanning Spectrophotometers
Toimittaja: Cole-Parmer
83056-22 83056-21 83056-23
634-1241EA 6750 EUR
634-1241 JENW83056-21 JENW83056-23
Cole-Parmer® SP-500 Visible and UV/Visible Scanning Spectrophotometers
Spektrofotometrit ja fotometrit UV/Vis-spektrofotometrit
Quick, easy navigation with large color touchscreen display.
  • Make setup easy with the user-friendly 177,8 mm high-definition color touchscreen and interface
  • Choice of measurement modes let you handle a multitude of applications
  • Simple connectivity with multiple USB ports for data storage and printer connectivity
  • Three-year warranty
  • Material: White ABS plastic
  • Bandwidth: 5 nm
Designed with your convenience in mind, the SP-500 series of spectrophotometers provides accurate and reliable results in various applications from teaching, to industrialMore Product Information