Stoppers for Standard Cells, Hellma Analytics
Toimittaja: HELLMA
011001-VE572 011002-VE572 011103-VE1073
634-1145EA 23.2 EUR
634-1145 634-1146 HELL011103-VE1073
Stoppers for Standard Cells, Hellma Analytics
Tulpat PE/LDPE/HDPE
An assortment of stoppers for use with standard Hellma® cells.
  • Hellma® precision cells are 100% manufactured in Germany and are suitable for all common spectrophotometers
  • Hellma® precision cells are characterised by the highest precision in path length accuracy and parallelism
  • Hellma® precision cells can be easily cleaned and reused and thus remain in use for a long time
PTFE stoppers with fitting NS5 are designed for use with cuvette types: 110, 111, 114, 120 with 1 to 5 mm path length, and for cuvette type 404 with 1 to 10 mm path length.