Visible spectrophotometer, GENESYS™ 30
Toimittaja: Thermo Fisher Scientific
840-277300
634-0910EA 3630 EUR
634-0910
Visible spectrophotometer, GENESYS™ 30
Spektrofotometrit ja fotometrit UV/Vis-spektrofotometrit
This high precision visible spectrophotometer can perform sophisticated analyses for both educational or industrial applications.
  • Single cell with removable and washable sample compartment liner
  • Cell holders dedicated to standard cells, including test tubes
  • Snap-on thermal printer for 80 mm paper width
  • Thermo Scientific™ VISIONlite PC software (optional)
In the laboratory, technicians working in routine QA/QC applications will obtain quality results that can be reported as digital or printed data.