UV/Vis spectrophotometers, U-3900
Toimittaja: VWR Collection
2J2-0032
634-0754EA 18860 EUR
634-0754
UV/Vis spectrophotometers, U-3900
Spektrofotometrit ja fotometrit UV/Vis-spektrofotometrit
U-3900 and U-3900H spectrophotometers are comprehensive, research-grade solutions. They provide excellent performance afforded by low stray light, selectable band pass and high energy.
  • Double-beam stability provides improved accuracy and repeatability over extended periods of time
  • Ability to reference-beam attenuate highly absorbing species for improved signal to noise
  • Selectable scanning rate range of 1,5 to 2400 nm/min and changeable scan rate for the UV region to minimise noise further
  • Interface via USB and a laptop or notebook PC format, can