Slide boxes, SlideFile™ Junior
Toimittaja: Simport Scientific
M700-50B M700-50G M700-50P M700-50W M700-50Y
631-1905EA 152 EUR
631-1905 631-1906 631-1907 631-1908 631-1909
Slide boxes, SlideFile™ Junior
Aluslasikotelot
Boxes are impact-resistant and stackable for space efficient storage.
  • Barcodes can be read through the clear lid
  • Number of slides can be doubled by storing two in a slot
  • Tinted hinged cover makes the content easy to see
Tilaustiedot: Include a slide box and a removable tray.