Peitinlasit, Menzel-Gläser
Toimittaja: Epredia
BB024036A113 BB021026A113 BB022032A113 BB024032A113 BB024040A113 BB024060A113 BC026076A140 BB024032AC13 BB024050A233 BB024060AC13 BC026076AC40 BB022050A113 BB024050AC13 BC050075A140 BB024055A113 BB024050A033 BB024060A033 BB022060A113 BB022032AC13 BB022040AC13 BB022060AC13 BB024036AC13 BB024060A233 BC040060A1 BC040050A1
631-0712EA 79.6 EUR
631-0712 631-1335 631-0852 631-0711 631-1333 631-1339 630-2910 630-2873 630-2867 631-0853 MENZBC026076AC40 631-1365 631-9430 631-9431 631-9444 630-2868 630-2864 630-2103 630-2879 631-1370 630-2877 630-2871 630-2895 MENZBC040060A1 MENZBC040050A1
Peitinlasit, Menzel-Gläser
Suojalasit
These cover slips are suitable for in vitro diagnostic (IVD) applications.
  • Uniform quality of thickness
  • No bubbles or flaws
  • Made of pre-washed pure white Hydrolytic Class 1 glass
Paksuus: 0,13 - 0,16 mm