Counting chambers, Malassez
Toimittaja: MARIENFELD
0640610 0640630
631-0975EA 47.8 EUR
631-0975 MARI0640630
Counting chambers, Malassez
Alus- ja peitinlasit Laskukammiot
Counting chambers specially designed for in vitro diagnostic.
  • Double net ruling
  • Two cover glasses
  • Dark lined without clamps