Paraffin embedding centres, modular, MPS/P2
Toimittaja: Slee
10170000 10172000 10177000 10180000 10182000 10185000 10187000 10175000
631-0574EA 9140 EUR
631-0574 631-0576 631-0579 SLEE10180000 631-0582 631-0583 631-0585 SLEE10175000
Paraffin embedding centres, modular, MPS/P2
Laitteet kudosten valamiseen
The MPS/P2 features ease of operation with ergonomic hand rest, spacious work surfaces, large container capacity and reliability. The extendable modular 3-component system with separate temperature control elements consists of a dispensing element MPS/P, a preheating unit MPS/W and a separate cooling plate MPS/C. The paraffin dispensing unit includes a 5,5 litre paraffin container. All three modules operate at the same working height and can be arranged according to laboratory requirements.
  • LED-illuminated working area
  • Adjustable forceps holders