Laboratory marker
Toimittaja: MARIENFELD
6130603
631-0014EA 5.3 EUR
631-0014
Laboratory marker
Kynät Lab Markers
Permanent marker for indelibly labelling microscope slides and embedding cassettes with a high contrast black ink. Ultra-fine tip. These laboratory markers do not dry out and have a long lifetime.
  • Water resistant, odour-free and non toxic
  • Resistant against most laboratory chemicals
  • No pretreatment of surfaces necessary