Digital USB cameras, Moticam® A
Toimittaja: MOTIC
1100600101412 1100600101192 1100600101302 1100600101443 1100600101463
630-3471EA 245 EUR
630-3471 630-3469 630-3470 630-3472 630-3473
Digital USB cameras, Moticam® A
Kamerat
Starter line sCMOS USB Camera Moticam® A for microscopy. All-In-One solution.
  • Different resolutions to fit to all needs and budgets
  • Immediately compatible with monocular and binocular microscopes thanks to the All-In-One solution, 2 eyepiece adaptors included
  • Macro tube for pictures in macro
  • MotiConnect Easy - flexible - downloadable software, easy to use with an intuitive interface
  • Driver UVC 'Universal Video Controller' self-installable, connects and installs itself
  • Automatic white balance
  • FPS up to 30 fps on full resolution
  • 2 years warranty