Upright microscope, DM750
Toimittaja: LEICA MICROSYSTEMS
13613010_P1 13613010_P2 13613010_P3 13613010_P4 13613010_P6 13613010_P7 13613010_P8
630-3277EA 2560 EUR
630-3277 630-3278 630-3279 630-3280 630-3282 630-3283 630-3284
Upright microscope, DM750
Mikroskoopit
Leica DM750 microscopes have been specifically developed to facilitate science teaching. DM750 is designed essentially for the versatile needs of advanced life science courses and for professional training, such as in medical, veterinary and dental schools.
  • Delivers high quality images of the most demanding specimens
  • Easy sample handling
  • Special sample holders with a 25 or 30 mm sample diameters
  • Stands with both four- or five-position nosepiece allow for a full range of magnifications between 50× and 1000×
  • Reflected light illuminator with cutting edge LED