USB camera, Moticam® S6
Toimittaja: MOTIC
1100600101111
630-3102EA 778 EUR
630-3102
USB camera, Moticam® S6
Kamerat
High Grade Moticam® S6 sCMOS USB microscopy camera for ultimate imaging result.
  • Fast frame rates
  • sCMOS sensors back-illuminated sensor technology
  • USB 3.1 interface
  • Motic Images Plus 3.0 software
The 6 MP resolution of Moticam S6 is targeted to large sized monitors and printing purposes. Presentations to a large audience through video projector and reports in hard copy are performed with ease. The price/performance ratio is also remarkable.
Tilaustiedot: To be purchased separately – C-mount adapter not included, this needs to be from the same brandMore Product Information