USB camera, Moticam® Pro S5 Lite
Toimittaja: MOTIC
1100600101101
630-3101EA 1350 EUR
630-3101
USB camera, Moticam® Pro S5 Lite
Kamerat
High Grade sCMOS USB Camera Moticam® Pro S5 Lite for microscopy for an ultimate imaging result for lowlight situations in Fluorescence and Polarization.
  • Fast frame rates
  • sCMOS sensors back-illuminated sensor technology
  • USB 3.1 interface
  • Ideal for fluorescence applications
  • Motic Images Plus 3.0 software
Low Light, Fast Phenomena – Moticam Pro S5 Lite delivers a 5MP mid-size resolution for all biological/medical fields, showing a premium sensitivity for lowlight situations in Fluorescence and Polarization. The Global shutter data readout helps toMore Product Information