VWR® VisiCam® PRO 20C, Digital Camera for Fluorescence Applications
Toimittaja: VWR Collection
630-2992EA 1870 EUR
630-2992
VWR® VisiCam® PRO 20C, Digital Camera for Fluorescence Applications
Kamerat
This high resolution C-mount camera is equipped with a 20 MP colour CMOS 1" sensor. The large sensor and cooling system make it ideal for low light applications like fluorescence.
  • Two-stage Peltier cooling system reaches −42 °C below ambient temperature: Increases the signal to noise ratio and decreases the image noise
  • Heat radiation is efficiently disposed of through electric fans to avoid moisture problems
  • USB 3.0 output ensures high speed data transmission
  • 'WaveImage' Professional software (included) for previewing and capturing images for later image