VWR®, Immersioöljy
Toimittaja: VWR Collection
630-1808EA 28 EUR
630-1808
VWR®, Immersioöljy
Värit Microscopy and staining reagents
Immersion oil is used to increase microscope resolution.
  • For immersing of the lens and specimen
  • Transparent oil with high refractive index
  • Increases the numerical aperture of an objective