Litiumsulfaattimonohydraatii, dried ≥99.0% (dried basis, T), Puriss. p.a. ACS, Fluka™

Toimittaja: Honeywell Chemicals
Varoitus

62609-100G 62609-500G
62609-100GEA 55 EUR
62609-100G 62609-500G
Litiumsulfaattimonohydraatii, dried ≥99.0% (dried basis, T), Puriss. p.a. ACS, Fluka™
Litiumsulfaattimonohydraatii
Kaava: Li₂SO₄·H₂O
Sulamispiste: 120 °C
Tiheys: 2,060 g/cm³
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149766
CAS-numero: 10102-25-7
EINECS: 233-820-4

Order Now

Testitulosten erittely

Assay (excluding dried substance) 99.0 - 101.0 %
Water insoluble matter Max. 0.01 %
Loss on drying (150°C) 13.0 - 15.0 %
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Heavy metals (as Pb) Max. 0.001 %
Chloride (Cl) Max. 20 ppm
Nitrate (NO₃) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR