VWR® TraceableGO, Temperature and Humidity Data Loggers with Bluetooth
Toimittaja: VWR Collection
620-2673EA 92.1 EUR
620-2673 620-2674
VWR® TraceableGO, Temperature and Humidity Data Loggers with Bluetooth
Dataloggerit
TraceableGO Bluetooth temperature and humidity dataloggers fill in the unknowns between destinations. Bluetooth device is the ideal transit partner for monitoring materials between destinations. Safely transport reagents, clinical samples, vaccines, pharmaceuticals and food items to any location.
  • Compact and easy to use
  • Critical information (temperature, trip duration, MIN/MAX, battery level and alarms) all displayed on LCD at all times
  • Compatible with TraceableGO or TraceableLIVE mobile apps
  • Devices store up to 90 alarm events, records readings once every