Water activity meter, AwTherm
Toimittaja: ROTRONIC INSTRUMENTS
AWTHERM-PS14 AWTHERM-PS40
620-2506EA 12320 EUR
620-2506 620-2507
Water activity meter, AwTherm
Hygrometrit Digitaaliset lämpö- ja kosteusmittarit
AwTherm is a temperature controlled water activity station – used for determining the water activity of a sample, maintained at a known temperature. Ideal to measure water activity in foodstuffs, cosmetics or pharmaceutical industries.
  • Sensor: HYGROMER® IN-1
  • Highest precision through exact stabilization of temperature
  • Wide temperature control range
  • Excellent reproducibility and long-term stability
  • Variable size of sample container
  • Reference probe interchangeable for calibration or cleaning
  • Fast measurements with the AwQuick function
  • Simple operation via