ASTM-Like precision thermometers, ACCU-SAFE
Toimittaja: Ludwig Schneider GmbH & CO. KG
1205086S 1205097S 1205085S 1205098S 1205130S 1205067S 1205009S 1205083S 1205019S 1205017S 1205021S 1205022S 1205001S 1202130S 1205082S 1205066S 1205065S 1205064S 1205020S 1205062S 1205084S 1205059S 1205058S 1205057S 1205063S 1202090S 1202097S 1202039S 1202057S 1202059S 1202040S 1202038S 1202037S 1202022S 1202021S 1202042S 1202083S 1202091S 1202089S 1202086S 1202058S 1202084S 1202082S 1202067S 1202066S 1202065S 1202064S 1202063S 1202062S 1202085S 1202020S 1202098S 1202017S 1202012S 1202009S 1202001S 1202019S
SCNL1205086SEA 125 EUR
SCNL1205086S SCNL1205097S SCNL1205085S SCNL1205098S SCNL1205130S SCNL1205067S SCNL1205009S SCNL1205083S SCNL1205019S SCNL1205017S SCNL1205021S SCNL1205022S SCNL1205001S 620-2497 SCNL1205082S SCNL1205066S SCNL1205065S SCNL1205064S SCNL1205020S SCNL1205062S SCNL1205084S SCNL1205059S SCNL1205058S SCNL1205057S SCNL1205063S SCNL1202090S SCNL1202097S SCNL1202039S SCNL1202057S 620-2459 SCNL1202040S SCNL1202038S 620-2494 SCNL1202022S SCNL1202021S SCNL1202042S SCNL1202083S 620-2458 620-2496 SCNL1202086S SCNL1202058S SCNL1202084S SCNL1202082S SCNL1202067S SCNL1202066S SCNL1202065S 620-2462 620-2454 620-2495 SCNL1202085S SCNL1202020S SCNL1202098S 620-2456 620-2453 SCNL1202009S 620-2455 SCNL1202019S
ASTM-Like precision thermometers, ACCU-SAFE
Lämpömittarit Analogue Thermometers
Precision thermometer, ideal to measure precisely and reliably without mercury from the very start.
  • Round, white back capillary form
  • Suitable for calibration
Blue filling, stem type.
Sertifikaatit: Conform to ASTM standards.
Tilaustiedot: Available with works certificate or with DAkkS calibration certificate (optional).
Pakkaus: Packed individually in a square plastic sleeve.