VWR® Traceable®, Lämpömittari, pakastimiin ja jääkaappeihin
Toimittaja: VWR Collection
620-2211EA 269 EUR
620-2211 620-2212 620-2213 620-2214 620-2215 620-2216 620-2217
VWR® Traceable®, Lämpömittari, pakastimiin ja jääkaappeihin
Lämpömittarit Digital Thermometers
These digital thermometers are designed for monitoring temperatures in refrigerators, freezers, water baths, heating blocks and incubators. Bottle probes are used in monitoring storage conditions for vaccines or other important samples.
  • Fixed 1 minute logging interval
  • Stores up to 10 unique alarm events
  • Capable of storing over one million observations
  • Raw data on unit cannot be overwritten or modified
  • Status: Low battery, low memory, USB data transfer and active alarm state
Bottle probe in non toxic glycol/bullet sensor/stainless steel probe with 3More Product Information