VWR® Traceable®, Vedenpitävät lämpömittarit
Toimittaja: VWR Collection
620-2092EA 74.5 EUR
620-2092 620-2099
VWR® Traceable®, Vedenpitävät lämpömittarit
Lämpömittarit Digital Thermometers
This waterproof and shockproof thermometer is a safe replacement for any hazardous mercury instrument. The probe is resistant to most chemicals.
  • Waterproof construction for wet labs, wash-down areas and all field applications
  • Memory recalls maximum/minimum temperatures
  • Reading updates every second
Stainless steel probe (Lר: 203×3,5 mm).
Toimitustiedot: Supplied with probe, probe cover, silver oxide battery and Traceable® certificate.