VWR® Traceable®, Vedenpitävä lämpömittari
Toimittaja: VWR Collection
620-2091EA 56.7 EUR
620-2091
VWR® Traceable®, Vedenpitävä lämpömittari
Lämpömittarit Digital Thermometers
This piercing thermometer can measure the internal temperature of stored, frozen, or prepared food. For use in soil, brewing, food processing, refrigeration and pharmaceutical applications.
  • Waterproof construction allows it to be used in labs, plants, wash-down areas, and the field
  • Manufactured to be drop-proof from 1,5 m
  • Reading (°C/°F) is updated every second
Sertifikaatit: Calibration complies with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.
Toimitustiedot: Supplied with probe, silver-oxide battery and Traceable® certificate of calibration.