VWR®, Hygrometri, digitaalinen, lämpö-kosteusmittari
Toimittaja: VWR Collection
620-1952EA 263 EUR
620-1952
VWR®, Hygrometri, digitaalinen, lämpö-kosteusmittari
Hygrometrit Digitaaliset lämpö- ja kosteusmittarit
This accurate thermo-hygrometer with barometer features additional derived units: Dew point temperature, wet bulb temperature, moisture content and absolute humidity. It is suitable for air conditioning applications, ambient air monitoring, meteorology, living climate and air pressure measurement in EDP rooms, storage rooms, greenhouses and production rooms.
  • Maximum/minimum value memory for all measurements
  • High accuracy by means of digital-works calibration
  • Offset and slope correction for easy adjustment
  • Configurable display: Alternating values
  • 'HOLD'