Dataloggeri, opetuskäyttöön, USBLink, opetus
Toimittaja: FOURIER SYSTEMS
USBLINK
620-1795EA 0 EUR
620-1795
Dataloggeri, opetuskäyttöön, USBLink, opetus
Dataloggerit
The USBLink is a sensor interface, converting any computer into a science data logging learning station. The USBLink simple 'plug-and-play' unit is highly functional, featuring four-input interface, with an option for up to eight simultaneously connected sensors via USB to the computer. The unit offers automatic sensor recognition, fast sampling rate and sample streaming of up to 10,000 samples per second.
  • Simple to operate
  • Simulataneous data collection for up to eight sensors
  • Works with full range of Fourier sensors
  • Powered directly by any PC USB port