Lämpömittari, tarkkuuslämpömittarit, TFE 510
Toimittaja: EBRO
1340-5510 1340-5516
620-1388EA 296 EUR
620-1388 620-1389
Lämpömittari, tarkkuuslämpömittarit, TFE 510
Lämpömittarit Digital Thermometers
This precision thermometer is a handy, watertight temperature measurement device for a whole range of measurement and control applications in the laboratory and in industry.
  • A micro-processor controls the electronics
  • Maximum measurement precision and linearity over the entire measurement range
  • Power off automatically after two hours
  • Measuring sensor: thermoelement
  • Three times faster than comparable RTD sensor based thermometers
Thermocouple type T sensor.
Toimitustiedot: Supplied with sensor (620-1389 only), a 3 V CR Lithium battery for up to 5More Product Information