Piirturi, lämpötilapiirturit, SC3
Toimittaja: DICKSON
SC377 SC386 SC387 SC367
620-0256EA 570 EUR
620-0256 620-0257 620-0258 620-0255
Piirturi, lämpötilapiirturit, SC3
Piirturit Piirturit
These recorders are used to record temperature simply and reliably. They provide the users with immediate access to data and suit applications in manufacture of pharmaceutical, medical and food products and vaccine storage.
  • Withstand the rigours of transport and storage
  • Accuracy and resolution conform to quality controls
  • For recordings over 24 hours or seven days
W×D×H: 93×93×57 mm
Weight: 227 mm
Toimitustiedot: Supplied with a single AA battery with a service life of two years and a red pen. Other diagrams are available on request.