Active pirani gauges
Toimittaja: EDWARDS HIGH VACUUM METTLER
D02171000 D02173000
619-0220EA 0 EUR
619-0220 619-0221
Active pirani gauges
Pumput Vakuumimittarit
Model APG.
These gauges have a measuring range from atmosphere to 1×10⁻⁴ mbar and combine the gauge head and controller in one compact active unit.
  • Wide range, regulated, internal power supply - runs from standard DC power supplies 20 to 36 V
  • Tolerant to voltage fluctuations
  • Medium and low-pressure versions available - APG-M and -MP from atmosphere to 10⁻³ mbar. APG-L from atmosphere to 10⁻⁴ mbar
  • Standard analogue output 0 to 10 V - Easy to interface with a computer or plc. Fault output indication
  • Adjustable set-point for process control and interlocking