Kloorivetyliuos 25%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Toimittaja: Avantor
Vaara

Synonyms: Hydrochloric acid solution, Muriatic acid

6167.2500EA 62.92 EUR
6167.2500 6167.9025
Kloorivetyliuos 25%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Kloorivetyliuos
Kaava: HCl
MW: 36.46 g/mol
Sulamispiste: –30 °C
Tiheys: 1.18 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011324
CAS-numero: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II

Order Now

Testitulosten erittely

Assay min. 25 %
Ammonium (NH4) max. 3 ppm
Free Chlorine max. 0.5 ppm
Residue after Ignition max. 5 ppm
Sulfate (SO4) max. 1 ppm
Sulfite (SO3) max. 2 ppm
Trace Impurities (in ppm)
Silver (Ag) max. 0.1
Aluminium (Al) max. 0.5
Arsenic (As) max. 0.1
Barium (Ba) max. 0.1
Beryllium (Be) max. 0.1
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.1
Cobalt (Co) max. 0.1
Chromium (Cr) max. 0.1
Copper (Cu) max. 0.1
Iron (Fe) max. 0.5
Germanium (Ge) max. 0.1
Potassium (K) max. 0.1
Lithium (Li) max. 0.1
Magnesium (Mg) max. 0.5
Manganese (Mn) max. 0.1
Molybdenum (Mo) max. 0.1
Sodium (Na) max. 0.5
Nickel (Ni) max. 0.1
Lead (Pb) max. 0.1
Strontium (Sr) max. 0.1
Titanium (Ti) max. 0.1
Thallium (Tl) max. 0.1
Vanadium (V) max. 0.1
Zinc (Zn) max. 0.5
Zirconium (Zr) max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR