Tiheyspullo, kalibroimaton
Toimittaja: Brand
43205 43208 43220 43228 43238
614-0339EA 90 EUR
614-0339 614-0341 614-0342 614-0343 614-0344
Tiheyspullo, kalibroimaton
Pyknometrit
Borosilicate 3.3 glass, Gay-Lussac type
  • Stopper NS 10/19 with capillary
  • Top of stopper ground and polished
  • Nominal capacity printed on the bottom
Sertifikaatit: DIN ISO 3507