Ominaispainomittari, tarkat ominaispainomittarit öljyteollisuuteen
Toimittaja: Alla France
3508MC07715BQP 3503LC07515BQP 3503LC08015BQP 3503LC08515BQP 3503LC09015B 3503LC09515BQP 3503MC07015BQP 3503MC07515BQP 3503LC06015BQP 3503LC07015BQP 3503LC10015BQP 3503LC10515BQP 3503MC06020BQP 3503MC06015BQP 3503MC06520BQP 3503MC06515BQP 3503MC07020BQP 3503MC07520BQP 3503MC08020BQP 3503MC08520BQP 3503MC09020BQP 3503MC9520BQP 3503MC10020BQP 3503MC10520BQP 3503SC07020BQP 3503LC08515EUC 3503MC06020EUC 3503MC06015EUC 3503MC06515EUC 3503MC07015EUC 3503MC07515EUC 3503MC08015EUC 3503MC08515EUC 3503MC09015EUC 3503MC09515EUC 3503MC10015EUC 3503MC10515EUC 3503LC09015EUC
614-0307EA 118 EUR
614-0307 614-0306 614-0308 614-0324 614-0319 614-0321 614-0304 614-0303 614-0299 614-0302 614-0322 614-0323 614-0328 614-0300 614-0331 614-0301 614-0325 614-0326 614-0332 614-0333 614-0334 614-0337 614-0335 614-0336 614-0338 ALLA3503LC08515EUC 614-0329 ALLA3503MC06015EUC ALLA3503MC06515EUC ALLA3503MC07015EUC ALLA3503MC07515EUC ALLA3503MC08015EUC ALLA3503MC08515EUC ALLA3503MC09015EUC ALLA3503MC09515EUC ALLA3503MC10015EUC ALLA3503MC10515EUC ALLA3503LC09015EUC
Ominaispainomittari, tarkat ominaispainomittarit öljyteollisuuteen
Ominaispainomittarit
These accurate hydrometers have been designed to meet the specific needs of users in the petroleum industry.
  • Reading below meniscus: Marking engraved on the stem to show any displacement of the scale
  • Low surface tension (16 to 35 mN/m)
  • Marked with the calibration temperature, surface tension, individual number of instrument and serial number
Sertifikaatit: ISO 649
Toimitustiedot: On request hydrometers can be supplied with EU traceable calibration certificate or EO official calibration certificate.