Sediment testers for milk
Toimittaja: GERBER FUNKE
4800 4810
511-0076EA 405 EUR
511-0076 614-0196
Sediment testers for milk
Suodatintelineet
SEDILAB: Manual sediment tester. With clamp for attaching to tables.
SEDILAB-E: Pump design sediment tester for serial testing of liquids for particle contamination, particularly for sediment testing of milk. Splash-proof design. Approx. 800 samples per hour. Sharply defined sediment images.
Pump design sediment tester for serial testing of liquids for particle contamination, particularly for sediment testing of milk.