Shakers
Toimittaja: GERBER FUNKE
3851 3850 3852
614-0184EA 2390 EUR
614-0184 614-0183 614-0185
Shakers
Ravistelijat Koeputkiravistelijat
Shakers for determining the fat content in milk according to Mojonnier's method. White PVC housing. Choice of models to accept 4, 6 or 36 Mojonnier extraction tubes. Units provide forceful, uniform and reproducible mixing.
  • Timer, 0 to 15 min
  • Produce 100 shakes per minute
  • Model for 36 butyrometers is supplied with stand