VWR® Reagent Reservoirs, PP
Toimittaja: VWR Collection
613-7288EA 290 EUR
613-7288 613-7295 613-7296 613-7302 613-7303 613-7313 613-7315 613-7316
VWR® Reagent Reservoirs, PP
Reagenssisäiliöt
These polypropylene multi-channel reagent reservoirs are designed to support both automated liquid handling platforms and manual systems. The reservoir design complies with ANSI/SLAS 1-2004, when used for cell culture and immunoassay experiments.
  • Multiple capacities and well numbers can be selected for different experimental needs
  • Multi-channel reagent reservoirs are suitable for both 8-channel and 12-channel pipettes
  • Rhombic well series: 96- or 384-well reagent reservoirs on the bottom
  • Non-sterile or sterilised by irradiation, SAL 10⁻⁶ (ISO 11137)
  • Lot-