VWR®, Single-Channel Pipettes, Mechanical, Variable Volume
Toimittaja: VWR Collection
613-2896EA 230 EUR
613-2896 613-2898 613-7102 613-7103 613-7104 613-7105 613-7106 613-7107 613-7108 613-7109 613-7110
VWR®, Single-Channel Pipettes, Mechanical, Variable Volume
Pipetit ja pipetintäyttäjät Pipetit
These lightweight Standard Line pipettes provide accuracy and precision. Their ergonomic features, such as the contoured shape of the handle and the soft spring system, deliver smooth and effortless pipetting, even during extensive periods of use.
  • Easily adjustable ejector height system to accommodate virtually all brands of tips
  • UV resistant and fully autoclavable
  • Easy and quick recalibration system
  • Colour coded shaft for easy volume identification
  • 5000 and 10000 μl version with shaft protected by filter