VWR®, Macro Pipette Tips
Toimittaja: VWR Collection
613-7094EA 29.2 EUR
613-7094 732-3912 613-7095 732-3913 613-7096 732-3914 613-7097 732-3915 613-7098 613-7099 613-7100 732-3916 613-7101 732-3917
VWR®, Macro Pipette Tips
Pipetinkärjet
Improved accuracy over serological pipettes.
  • Tip length facilitates access into bottles, cell culture flasks, and tubes
Universal fit for compatibility with most major pipettor brands.
Sertifikaatit: Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free. Sterile product is certified pyrogen-free.