Bottle top tilt measures
Toimittaja: Lenz Laborglas GmbH & CO.KG
05476010 05476019 05476028 05476022 05476037 05476001 05476007 05476013 05476004
613-6256EA 74.9 EUR
613-6256 613-6252 613-6255 613-0696 613-0697 613-6251 613-6254 613-6250 613-6253
Bottle top tilt measures
Pullon kallistuksen mittauslaitteet
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
This dispenser head is completed by a 1 litre bottle for repetitive dispensing of the same volume.
NS 29/32