Pulloannostelija, Calibrex™ Solutae 530
Toimittaja: Socorex Isba
530.025 530.050 530.100 530.025FC 530.050FC 530.100FC 530.001 530.001FC 530.002.5 530.002.5FC 530.005 530.005FC 530.010 530.010FC
613-5990EA 648 EUR
613-5990 613-5992 613-5994 613-5991 613-5993 613-5995 613-5982 613-5983 613-5984 613-5985 613-5986 613-5987 613-5988 613-5989
Pulloannostelija, Calibrex™ Solutae 530
Pulloannostelijat
The dispensers have a PFA coated preventing plunger and barrel from seising together. This enables trouble free distribution of salt solutions, weak and strong acids, as well as bases.
  • Glass or ceramic PFA coated plunger for salt solutions, weak and strong acids, as well as bases
  • QR coded chemical compatibility
  • Integrated calibration key
  • Quick disassembling, no tool required
  • Autoclavable at 121 °C fully assembled
Flow control stopcok available, for priming without reagent loss or contamination, with locking position for safe dispenser transportation.More Product Information