Capillary micropipettes, MICROMAN® E
Toimittaja: GILSON
FD10001 FD10006 FD10005 FD10004 FD10002 FD10003
613-5825EA 349 EUR
613-5825 613-5830 613-5829 613-5828 613-5826 613-5827
Capillary micropipettes, MICROMAN® E
Pipetit ja pipetintäyttäjät Pipetit
The ergonomically designed MICROMAN® E positive-displacement pipettes, along with disposable capillary piston (CP) tips, provide the highest precision when pipetting viscous, non-aqueous liquids such as oils, blood, glycerol, and more while protecting the pipette from contamination.
  • Reliable results even with non-aqueous liquids
  • Overall protection
  • Designed with comfort in mind
Contaminated, corrosive, or radioactive samples can generate aerosols that can cross-contaminate or damage the pipette. The disposable piston within the capillary piston (CP)More Product Information