Monikanavaiset pipetit, mekaaniset, Finnpipette™ F1
Toimittaja: Thermo Fisher Scientific
4661000N 4661010N 4661020N 4661030N 4661040N 4661050N 4661060N 4661070N 4661080N 4661090N
613-5568EA 631 EUR
613-5568 613-5569 613-5570 613-5571 613-5572 613-5573 613-5574 613-5575 613-5576 613-5577
Monikanavaiset pipetit, mekaaniset, Finnpipette™ F1
Pipetit ja pipetintäyttäjät Pipetit
Finnpipette™ F1 pipettes deliver performance with outstanding precision in a lightweight design that maximises user comfort and reduces strain.
  • For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide
  • Silent and lighter volume adjustment mechanism with over 50% lower forces compared to earlier models
  • Improved grip on the volume adjustment knob ensures effortless and precise adjustment
  • Secure volume setting prevents undesirable volume drift during