Yksikanavaiset pipetit, mekaaniset, säädettävä tai kiinteä tilavuus, Finnpipette™ F1
Toimittaja: Thermo Fisher Scientific
4641010N 4641020N 4641030N 4641040N 4641050N 4641060N 4641070N 4641080N 4641090N 4641100N 4641110N 4641120N 4641130N 4641140N 4651020N 4651030N 4651040N 4651050N 4651060N 4651070N 4651080N 4651090N 4651100N 4651110N 4651120N 4651130N 4651140N
613-5541EA 238 EUR
613-5541 613-5542 613-5543 613-5544 613-5545 613-5546 613-5547 613-5548 613-5549 613-5550 613-5551 613-5552 613-5553 613-5554 613-5555 613-5556 613-5557 613-5558 613-5559 613-5560 613-5561 613-5562 613-5563 613-5564 613-5566 613-5565 613-5567
Yksikanavaiset pipetit, mekaaniset, säädettävä tai kiinteä tilavuus, Finnpipette™ F1
Pipetit ja pipetintäyttäjät Pipetit
Finnpipette™ F1 pipettes deliver performance with outstanding precision in a lightweight design that maximises user comfort and reduces strain.
  • Silent and lighter volume adjustment mechanism with over 50% lower forces compared to earlier models
  • Improved grip on the volume adjustment knob ensures effortless and precise adjustment
  • Secure volume setting prevents undesirable volume drift during operation
  • 120° adjustable finger rest - for both right- and left-handed operator comfort - allows the hand to relax during pipetting cycles
  • Lightweight handle design