VWR®, Multi-Channel Pipettes, Electronic, Variable Volume
Toimittaja: VWR Collection
613-5419EA 1150 EUR
613-5419 613-5420 613-5421 613-5422 613-5423 613-5424 613-5425 613-5426 613-5782 613-5783
VWR®, Multi-Channel Pipettes, Electronic, Variable Volume
Pipetit ja pipetintäyttäjät Pipetit
These pipettes combine superior accuracy, precision and robustness with advanced ergonomic design.
  • Ease of use: Only two buttons and five pipetting modes; enables an easy one-handed programming and a quick pipetting cycle start
  • CUSTOM mode: Users can quickly and easily create specific protocols thanks to VWR® Protocol Builder software
  • Advanced ergonomics: Lightweight and perfectly balanced, it requires virtually no force to significantly reduce RSI
  • Fits comfortably in either the left or right hand and allows a 360° rotating manifold
  • Fully motorised piston